Learn More
Medchemexpress LLC 2-[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methylsulfonyl]-1H-benzimidazole | 117976-47-3 | MFCD08063743 | 100mg
SDP
Catalog No. 5000267761
Encompass_Preferred
Rabeprazole sulfone is a metabolite of the proton pump inhibitor Rabeprazole and is an impurity reference for Rabeprazole[1]
Catalog No. 50-002-67761 Supplier Medchemexpress LLC Supplier No. HYZ2337100MG
Rabeprazole sulfone is a metabolite of the proton pump inhibitor Rabeprazole and is an impurity reference for Rabeprazole[1]