Learn More
Strem, An Ascensus Company CAS# 480424-68-8. 5g. 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate, min. 97%. MFCD03453062
SDP
Catalog No. 5090108746
Encompass
CAS# 480424-68-8. 5g. 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate, min. 97%. MFCD03453062. Molecular Weight: 262.14. Molecular Formula: C14H19BO4. Color/form: colorless xtl. Strem# 05-1045. http://www.strem.com/catalog/v/05-1045/
Catalog No. 50-901-08746 Supplier Strem, An Ascensus Company Supplier No. 0510455G
CAS# 480424-68-8. 5g. 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate, min. 97%. MFCD03453062. Molecular Weight: 262.14. Molecular Formula: C14H19BO4. Color/form: colorless xtl. Strem# 05-1045. http://www.strem.com/catalog/v/05-1045/