Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[4-(4,5-diphenyl-1H-imidazol-2-yl)phenyl]-4,5-diphenyl-1H-imidazole | 4051-59-6 | 98.0% | 514.62 g/mol | C36H26N4 | 5 MG
SDP

Catalog No. 5000247389
Encompass_Preferred

MSX-130 is a small-molecule CXCR4 antagonist used in research to study cancer metastasis. It is supplied as a high-purity solid for laboratory use and is intended for in vitro and in vivo research applications; not for human or clinical use.

  • Research reagent for CXCR4 antagonism.
  • High purity: 98.0%.
  • Molecular weight: 514.62 g/mol.
  • Molecular formula: C36H26N4.
  • Supplied as a pre-weighed solid for convenient dosing.
  • Suitable for cellular and animal model studies.

Catalog No. 50-002-47389 Supplier Medchemexpress LLC Supplier No. HY1030105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MSX-130 is a small-molecule CXCR4 antagonist used in research to study cancer metastasis. It is supplied as a high-purity solid for laboratory use and is intended for in vitro and in vivo research applications; not for human or clinical use.

  • Research reagent for CXCR4 antagonism.
  • High purity: 98.0%.
  • Molecular weight: 514.62 g/mol.
  • Molecular formula: C36H26N4.
  • Supplied as a pre-weighed solid for convenient dosing.
  • Suitable for cellular and animal model studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.