Learn More
Medchemexpress LLC 2-(4-(4-(Aminomethyl)-1-oxo-1,2-dihydrophthalazin-6-yl)-1-methyl-1H-pyrazol-5-yl)-3-fluoro-1-naphthonitrile hydrochloride | 99.88% | 460.89 | 25 MG
SDP
Catalog No. 5000348485
Encompass_Preferred
MRTX9768 hydrochloride is a potent, selective, orally active, first-in-class PRMT5-MTA complex inhibitor.
  • Potent, selective, orally active, first-in-class PRMT5-MTA complex inhibitor.
  • Inhibits SDMA and cell proliferation in HCT116 MTAP-del cells.
  • Selectively targets MTAP/CDKN2A-deleted tumors, such as glioblastoma.
  • Demonstrates dose-dependent inhibition of SDMA in MTAP-del tumors in xenograft studies.
  • Exhibits a favorable ADME profile with good bioavailability in mice and dogs.
Catalog No. 50-003-48485 Supplier Medchemexpress LLC Supplier No. 25MGHY138684A
MRTX9768 hydrochloride is a potent, selective, orally active, first-in-class PRMT5-MTA complex inhibitor.
  • Potent, selective, orally active, first-in-class PRMT5-MTA complex inhibitor.
  • Inhibits SDMA and cell proliferation in HCT116 MTAP-del cells.
  • Selectively targets MTAP/CDKN2A-deleted tumors, such as glioblastoma.
  • Demonstrates dose-dependent inhibition of SDMA in MTAP-del tumors in xenograft studies.
  • Exhibits a favorable ADME profile with good bioavailability in mice and dogs.