Learn More
Medchemexpress LLC 2-[[4-(4-chlorophenyl)-1-piperazinyl]methyl]-pyrazolo[1,5-a]pyridine | 337972-47-1 | MFCD06798338 | 99.8% | C18H19ClN4 | 10MG
SDP
Catalog No. 5000259676
Encompass_Preferred
FAUC 213 is an orally active, highly selective dopamine D4 receptor complete antagonist used as a research compound in neuroscience and pharmacology studies. It shows high affinity for the human D4.4 receptor, reduced activity at D2 and D3 receptors, and can cross the blood-brain barrier.
  • High affinity for dopamine D4 receptor (Ki = 2.2 nM for hD4.4).
  • Low activity at D2 and D3 receptors (Ki = 3.4 μM and 5.3 μM, respectively).
  • Orally active and brain penetrant.
  • Useful as a tool to study dopaminergic signaling and atypical antipsychotic profiles.
  • Research use only; not for human or clinical use.
Catalog No. 50-002-59676 Supplier Medchemexpress LLC Supplier No. HY1432710MG
FAUC 213 is an orally active, highly selective dopamine D4 receptor complete antagonist used as a research compound in neuroscience and pharmacology studies. It shows high affinity for the human D4.4 receptor, reduced activity at D2 and D3 receptors, and can cross the blood-brain barrier.
  • High affinity for dopamine D4 receptor (Ki = 2.2 nM for hD4.4).
  • Low activity at D2 and D3 receptors (Ki = 3.4 μM and 5.3 μM, respectively).
  • Orally active and brain penetrant.
  • Useful as a tool to study dopaminergic signaling and atypical antipsychotic profiles.
  • Research use only; not for human or clinical use.