Learn More
Medchemexpress LLC 2-[4-(4-chlorophenyl)piperazin-1-ylmethyl]pyrazolo[1,5-a]pyridine | 337972-47-1 | MFCD06798338 | 99.9% | 326.82 g/mol | C18H19ClN4 | 5 MG
SDP
Catalog No. 5000222116
Encompass_Preferred
FAUC 213 is a small-molecule research compound acting as a highly selective dopamine D4 receptor full antagonist. It is supplied as a solid for pharmacology studies and analytical use, with defined molecular properties and high reported purity suitable for reproducible in vitro and in vivo experiments.
  • Highly selective dopamine D4 receptor antagonist for receptor pharmacology studies.
  • High purity (~99.9%) for consistent experimental results.
  • Available in milligram-scale quantities to support screening and dose-ranging studies.
  • Well-characterized molecular formula and weight for formulation and calculations.
  • Offered as solid and as a DMSO solution option for assay-ready formats.
Catalog No. 50-002-22116 Supplier Medchemexpress LLC Supplier No. HY143275MG
FAUC 213 is a small-molecule research compound acting as a highly selective dopamine D4 receptor full antagonist. It is supplied as a solid for pharmacology studies and analytical use, with defined molecular properties and high reported purity suitable for reproducible in vitro and in vivo experiments.
  • Highly selective dopamine D4 receptor antagonist for receptor pharmacology studies.
  • High purity (~99.9%) for consistent experimental results.
  • Available in milligram-scale quantities to support screening and dose-ranging studies.
  • Well-characterized molecular formula and weight for formulation and calculations.
  • Offered as solid and as a DMSO solution option for assay-ready formats.