Learn More
Medchemexpress LLC 2-((4,4-dimethyl-1-oxidothiochroman-6-yl)ethynyl)pyrimidine-5-carboxylic acid | 2131803-93-3 | MFCD32708542 | 99.2% | C18H16N2O3S | 10MG
SDP
Catalog No. 5000261515
Encompass_Preferred
This tazarotene-derivative retinoid small molecule is provided for laboratory research. It contains an alkyne group for click-chemistry applications and is characterized for purity, solubility, and storage stability to support biochemical and cellular studies.
  • High purity suitable for research assays (approximately 99.2%).
  • Solid, light yellow to yellow powder for easy handling.
  • Alkyne functional group compatible with click chemistry workflows.
  • Soluble in DMSO at about 50 mg/mL; sonication recommended.
  • Stable under recommended storage: powder at -20°C (long term) and 4°C (short term).
  • Supplied in small research pack sizes to support assay development.
Catalog No. 50-002-61515 Supplier Medchemexpress LLC Supplier No. HY13655410MG
This tazarotene-derivative retinoid small molecule is provided for laboratory research. It contains an alkyne group for click-chemistry applications and is characterized for purity, solubility, and storage stability to support biochemical and cellular studies.
  • High purity suitable for research assays (approximately 99.2%).
  • Solid, light yellow to yellow powder for easy handling.
  • Alkyne functional group compatible with click chemistry workflows.
  • Soluble in DMSO at about 50 mg/mL; sonication recommended.
  • Stable under recommended storage: powder at -20°C (long term) and 4°C (short term).
  • Supplied in small research pack sizes to support assay development.