Learn More
Medchemexpress LLC 2-((4,4-dimethyl-2-methylenecyclopent-1-en-1-yl)amino)-4-phenyl-1,3-naphthyridine | 511514-03-7 | ≥98.0% | 343.42 g/mol | C21H21N5 | 5 MG
SDP
Catalog No. 5000247840
Encompass_Preferred
A small-molecule inhibitor that selectively binds the heterotrimeric Gαi subunit and blocks α2-adrenergic receptor-mediated regulation of intracellular cAMP. Intended for laboratory research use only.
  • Selective inhibition of the Gαi subunit, blocking α2AR-mediated cAMP regulation.
  • Small molecule with molecular formula C21H21N5 and molecular weight 343.42 g/mol.
  • High purity (≥98%) and light yellow to yellow solid appearance.
  • Soluble in DMSO (250 mg/mL); example in vivo formulations documented.
  • Stable as powder at -20°C for long-term storage; defined solvent stability at lower temperatures.
  • Supplied in a 5 mg quantity for laboratory testing.
Catalog No. 50-002-47840 Supplier Medchemexpress LLC Supplier No. HY1020765MG
A small-molecule inhibitor that selectively binds the heterotrimeric Gαi subunit and blocks α2-adrenergic receptor-mediated regulation of intracellular cAMP. Intended for laboratory research use only.
  • Selective inhibition of the Gαi subunit, blocking α2AR-mediated cAMP regulation.
  • Small molecule with molecular formula C21H21N5 and molecular weight 343.42 g/mol.
  • High purity (≥98%) and light yellow to yellow solid appearance.
  • Soluble in DMSO (250 mg/mL); example in vivo formulations documented.
  • Stable as powder at -20°C for long-term storage; defined solvent stability at lower temperatures.
  • Supplied in a 5 mg quantity for laboratory testing.