Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[4-[[4-hydroxy-3-[2-(4-nitrophenyl)ethynyl]-5-propan-2-ylphenyl]methyl]-3,5-dimethylphenoxy]aceti | 447415-26-1 | 99.4% | C28H27NO6 | 10MG
SDP

Catalog No. 5000258815
Encompass_Preferred

NH-3 is an orally active, reversible thyroid hormone receptor (THR) antagonist that inhibits hormone binding and cofactor recruitment; it shows potent THR inhibition (IC50 55 nM) and contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition for click chemistry applications.

  • Orally active, reversible thyroid hormone receptor antagonist.
  • Potent THR inhibition with reported IC50 of 55 nM.
  • Contains an alkyne group for copper-catalyzed click chemistry.
  • High purity suitable for research use (≈99.4%).
  • Available as powder and DMSO solution formats for flexibility in assays.
  • Recommended storage conditions provided to preserve stability.

Catalog No. 50-002-58815 Supplier Medchemexpress LLC Supplier No. HY14151310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-3 is an orally active, reversible thyroid hormone receptor (THR) antagonist that inhibits hormone binding and cofactor recruitment; it shows potent THR inhibition (IC50 55 nM) and contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition for click chemistry applications.

  • Orally active, reversible thyroid hormone receptor antagonist.
  • Potent THR inhibition with reported IC50 of 55 nM.
  • Contains an alkyne group for copper-catalyzed click chemistry.
  • High purity suitable for research use (≈99.4%).
  • Available as powder and DMSO solution formats for flexibility in assays.
  • Recommended storage conditions provided to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.