Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[4-[(4-methoxyphenyl)methoxy]-phenyl]acetonitrile | 1016535-83-3 | MFCD09929139 | 98.1% | 241.26 g/mol | C15H12FNO | 10 MG
SDP

Catalog No. 5000299030
Encompass_Preferred

Oct3/4-inducer-1 is a small-molecule activator that promotes expression, stabilization, and transcriptional activity of the Oct3/4 (Oct4) transcription factor in human somatic cells. Supplied for research use only, it is used in studies of pluripotency and cellular reprogramming.

  • Promotes expression and stabilization of Oct3/4.
  • Enhances Oct3/4 transcriptional activity in diverse human somatic cells.
  • Provided as a solid with high reported purity (98.1%).
  • Soluble in DMSO at 100 mg/mL for preparation of stock solutions.
  • Suitable for studies of pluripotency and induced pluripotent stem cell formation.
  • Recommended storage: solid at 4°C; aliquots in solution at -80°C for long-term storage.

Catalog No. 50-002-99030 Supplier Medchemexpress LLC Supplier No. HY1877310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oct3/4-inducer-1 is a small-molecule activator that promotes expression, stabilization, and transcriptional activity of the Oct3/4 (Oct4) transcription factor in human somatic cells. Supplied for research use only, it is used in studies of pluripotency and cellular reprogramming.

  • Promotes expression and stabilization of Oct3/4.
  • Enhances Oct3/4 transcriptional activity in diverse human somatic cells.
  • Provided as a solid with high reported purity (98.1%).
  • Soluble in DMSO at 100 mg/mL for preparation of stock solutions.
  • Suitable for studies of pluripotency and induced pluripotent stem cell formation.
  • Recommended storage: solid at 4°C; aliquots in solution at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.