Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4,4'-Trihydroxychalcone, 97%, Thermo Scientific™

Catalog No. p-7029456
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2692003 1 g
AAH2692006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2692003 Supplier Thermo Scientific Chemicals Supplier No. H2692003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 961-29-5
Molecular Formula C15H12O4
Molecular Weight (g/mol) 256.257
MDL Number MFCD00075907
InChI Key DXDRHHKMWQZJHT-FPYGCLRLSA-N
Synonym isoliquiritigenin, 2',4,4'-trihydroxychalcone, 4,2',4'-trihydroxychalcone, 6'-deoxychalcone, e-1-2,4-dihydroxyphenyl-3-4-hydroxyphenyl prop-2-en-1-one, 2',4',4-trihydroxychalcone, isoliquirtigenin, unii-b9cti9gb8f, ccris 7676, chalcone, 2',4,4'-trihydroxy
PubChem CID 638278
ChEBI CHEBI:310312
IUPAC Name (E)-1-(2,4-dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one
SMILES C1=CC(=CC=C1C=CC(=O)C2=C(C=C(C=C2)O)O)O

Specifications

Melting Point 185°C to 188°C
Quantity 1 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 256.27
Percent Purity 97%
Chemical Name or Material 2',4,4'-Trihydroxychalcone
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber FL7080000
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.