Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4,4-Trimethyl-1-pentene

Catalog No. 44301825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-301-825ML 25 mL
44-301-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-301-825ML Supplier Sigma Aldrich Supplier No. 44301825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4,4-trimethyl-1-pentene is a α-alkene
  • It is the main dimeric product of isobutene
  • Ozonolysis of 2,4,4-trimethyl-1-pentene has been investigated in a flow reactor under pseudo-first-order conditions
  • Hydrocracking (hcg) of 2,4,4-trimethyl-1-pentene has been employed as a model test reaction to investigate the catalytic functionalities of sulfided como catalysts
  • Liquid phase oxidation of 2,4,4-trimethyl-1-pentene with molecular xygen afforded epoxides and hydrogen peroxides

Specifications

CAS 107-39-1
Density 0.708 g/mL (at 25°C (literature))
Boiling Point 101°C to 102°C (lit.)
Molecular Formula C8H16
Refractive Index n20/D 1.408 (literature)
Linear Formula (CH3)3 CCH2C(CH3)=CH2
MDL Number MFCD00008855
Quantity 25 mL
Synonym alpha-Diisobutylene; Diisobutylene
Molecular Weight (g/mol) 112.21
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.