Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,4-Trimethyl-1-pentene, Mixture of Isomers 95%, Thermo Scientific™

Catalog No. AC407780010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407780010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407780010 Supplier Thermo Scientific Chemicals Supplier No. 407780010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-39-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

Acidity 0.001% max. (as H2SO4)
CAS Min % 94.0
CAS Max % 100.0
Density 0.72
Infrared Spectrum Authentic
Refractive Index 1.4080 to 1.4100 (20°C, 589nm)
Linear Formula CH2C(CH3)CH2C(CH3)3
Quantity 1 kg
Formula Weight 112.22
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 2,4,4-Trimethyl-1-pentene, mixture of isomers, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.