Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid

Catalog No. AAJ64414MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64414MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64414MA Supplier Thermo Scientific Chemicals Supplier No. J64414MA
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19231-60-8 | C10H3I4NO4 | 708.755 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19231-60-8
Molecular Formula C10H3I4NO4
Molecular Weight (g/mol) 708.755
MDL Number MFCD05023065
InChI Key FLSXICNYHDJIAW-UHFFFAOYSA-N
Synonym n,n-tetraiodo-phthaloyl-glycine, 2-4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl acetic acid, 2-4,5,6,7-tetraiodo-1,3-dioxoisoindolin-2-yl acetic acid, 4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl acetic acid, 4,5,6,7-tetraiodo-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-4,5,6,7-tetraiodo-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid
PubChem CID 4462449
IUPAC Name 2-(4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl)acetic acid
SMILES C(C(=O)O)N1C(=O)C2=C(C1=O)C(=C(C(=C2I)I)I)I

Specifications

Quantity 10 mg
Formula Weight 708.75
Physical Form Powder
Chemical Name or Material 2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.