Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichlorobenzenethiol 97.0+%, TCI America™
SDP

Catalog No. T072810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072810G 10 g
T072825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T072810G Supplier TCI America Supplier No. T072810G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3773-14-6
Molecular Formula C6H3Cl3S
Molecular Weight (g/mol) 213.5
MDL Number MFCD00014447
InChI Key JARIALSGFXECCH-UHFFFAOYSA-N
Synonym 2,4,5-trichlorothiophenol, renacit ii, benzenethiol, 2,4,5-trichloro, 2,4,5-trichlorobenzene-1-thiol, acmc-209iv9, 2,4,5-trichloro-thiophenol, 2,4,5-trichlorophenyl mercaptan, 2,4,5-tris chloranyl benzenethiol, n-memet-n-trimetsilylmet benzylamine
PubChem CID 19597
IUPAC Name 2,4,5-trichlorobenzenethiol
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 154°C
Quantity 10 g
Formula Weight 213.50
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichlorobenzenethiol
RTECSNumber DC1948000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.