Learn More
AdipoGen 2 4 5-Trichlorophenol
SDP
Supplier:  AdipoGen CDXT0112G025
Encompass_Preferred
Chemical. CAS 95-95-4. Formula C6H3Cl3O. MW 197.5. Synthetic. Building block for synthesis of pesticides. Used as a broad range pesticide against insects, fungi, vegetation and bacteria. It has become a common environmental contaminant and probable human carcinogen. Compound can be used as analytical reference material.
Catalog No. 50-256-0006