Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichlorophenyl Disulfide 98%, Thermo Scientific™

Catalog No. AC190450500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190450500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC190450500 Supplier Thermo Scientific Chemicals Supplier No. 190450500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3808-87-5
Molecular Formula C12H4Cl6S2
Molecular Weight (g/mol) 424.98
MDL Number MFCD00000551
InChI Key ZUVJVEOHQKNMPN-UHFFFAOYSA-N
Synonym bis 2,4,5-trichlorophenyl disulfide, 2,4,5-trichlorophenyl disulfide, unii-ee043utu8o, 1,2,4-trichloro-5-2,4,5-trichlorophenyl disulfanyl benzene, 2,4,5-trichlorophenyl disulphide, ee043utu8o, bis 2,4,5-trichlorophenyl disulphide, 1,1'-dithiobis 2,4,5-trichlorobenzene, acmc-1ajg6
PubChem CID 77421
IUPAC Name 1,2,4-trichloro-5-[(2,4,5-trichlorophenyl)disulfanyl]benzene
SMILES ClC1=CC(Cl)=C(SSC2=CC(Cl)=C(Cl)C=C2Cl)C=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Quantity 50 g
Linear Formula (Cl3C6H2)2S2
Formula Weight 425
Chemical Name or Material 2, 4, 5-Trichlorophenyl disulfide, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.