Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluoro-1,3-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. T20601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20601G 1 g
T20605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20601G Supplier TCI America Supplier No. T20601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 321182-37-0
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD04114298
InChI Key ONVSXVVJFJJZQX-UHFFFAOYSA-N
Synonym 1,3-Diamino-2,4,5-trifluorobenzene
PubChem CID 15531223
IUPAC Name 2,4,5-trifluorobenzene-1,3-diamine
SMILES C1=C(C(=C(C(=C1F)F)N)F)N

Specifications

Melting Point 76°C
Quantity 1 g
Formula Weight 162.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trifluoro-1,3-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.