Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4',5'-Trifluoroacetophenone, 99%, Thermo Scientific™

Catalog No. AAH2620506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2620506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2620506 Supplier Thermo Scientific Chemicals Supplier No. H2620506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129322-83-4
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00061193
InChI Key GVTLJUZWNNFHMZ-UHFFFAOYSA-N
Synonym 2',4',5'-trifluoroacetophenone, 1-2,4,5-trifluorophenyl ethanone, 2,4,5-trifluoroacetophenone, 1-2,4,5-trifluorophenyl ethan-1-one, ethanone, 1-2,4,5-trifluorophenyl, 1-2,4,5-trifluoro-phenyl-ethanone, 1-acetyl-2,4,5-trifluorobenzene, pubchem4288, acmc-209bgn, 2',4',5'-trifluoro acetophenone
PubChem CID 594417
IUPAC Name 1-(2,4,5-trifluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1F)F)F

Specifications

Density 1.331
Flash Point 71°C (160°F)
Refractive Index 1.4665
Quantity 5 g
Formula Weight 174.12
Percent Purity 99%
Chemical Name or Material 2',4',5'-Trifluoroacetophenone
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.