Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4',5'-Trifluoroacetophenone 97.0+%, TCI America™
SDP

Catalog No. T27585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27585G 5 g
T27581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27585G Supplier TCI America Supplier No. T27585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129322-83-4
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00061193
InChI Key GVTLJUZWNNFHMZ-UHFFFAOYSA-N
Synonym 2',4',5'-trifluoroacetophenone, 1-2,4,5-trifluorophenyl ethanone, 2,4,5-trifluoroacetophenone, 1-2,4,5-trifluorophenyl ethan-1-one, ethanone, 1-2,4,5-trifluorophenyl, 1-2,4,5-trifluoro-phenyl-ethanone, 1-acetyl-2,4,5-trifluorobenzene, pubchem4288, acmc-209bgn, 2',4',5'-trifluoro acetophenone
PubChem CID 594417
IUPAC Name 1-(2,4,5-trifluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1F)F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 174.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′,5′-Trifluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.