Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. T23195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23195G 5 g
T23191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23195G Supplier TCI America Supplier No. T23195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 165047-24-5
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.095
MDL Number MFCD00061196
InChI Key CYIFJRXFYSUBFW-UHFFFAOYSA-N
Synonym 2,4,5-trifluoro benzaldehyde, benzaldehyde, 2,4,5-trifluoro, benzaldehyde, 2,4,5-trifluoro-9ci, pubchem1468, pubchem4258, intermediates-zcf02238, acmc-209ds3, 2,4,5-trifluorbenzaldehyde, 2,4,5-trifluorobenzoaldehyde, ksc494i3d
PubChem CID 519278
IUPAC Name 2,4,5-trifluorobenzaldehyde
SMILES C1=C(C(=CC(=C1F)F)F)C=O

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 g
UN Number 1989
Formula Weight 160.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.