Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. T23865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23865G 5 g
T23861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23865G Supplier TCI America Supplier No. T23865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157911-56-3
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00061209
InChI Key GAUXEYCSWSMMFZ-UHFFFAOYSA-N
PubChem CID 2777057
IUPAC Name 1-(bromomethyl)-2,4,5-trifluorobenzene
SMILES C1=C(C(=CC(=C1F)F)F)CBr

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Synonym 2,4,5-trifluorobenzyl bromide, 1-bromomethyl-2,4,5-trifluorobenzene, 2,4,5-trifluorobenzylbromide, 2,4,5-trifluoro benzyl bromide, benzene, 1-bromomethyl-2,4,5-trifluoro, alpha-bromo-2,4,5-trifluorotoluene, 2, 4, 5-trifluorobenzyl bromide, pubchem3026, 1-bromomethyl-2,4,5-trifluoro-benzene, acmc-1c0kl
Formula Weight 225.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluorobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.