Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T28891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28891G 1 g
T28895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28891G Supplier TCI America Supplier No. T28891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 243139-71-1
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.55
MDL Number MFCD00236319
InChI Key JMXPOOVDUVHJRO-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobenzyl chloride, 1-chloromethyl-2,4,5-trifluorobenzene, 2,4,5-trifluorobenzylchloride, 2,4,5-trifluorobenzyl choride, benzene, 1-chloromethyl-2,4,5-trifluoro, alpha-chloro-2,4,5-trifluorotoluene, 1-chloromethyl-2,4,5-trifluoro-benzene, pubchem4024, acmc-1cjlx
PubChem CID 2777063
IUPAC Name 1-(chloromethyl)-2,4,5-trifluorobenzene
SMILES FC1=CC(F)=C(CCl)C=C1F

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 180.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.