Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorotoluene, 98%, Thermo Scientific™

Catalog No. AAH6319003
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AAH6319003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6319003 Supplier Thermo Scientific Chemicals Supplier No. H6319003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 887267-34-7
Molecular Formula C7H5F3
Quantity 1g
MDL Number MFCD08458011
Synonym 2,4,5-trifluorotoluene, pubchem9128, 1,2,4-trifluoro-5-methyl-benzene
InChI Key ZGEAYXBIJAYBKA-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1F)F)F
IUPAC Name 1,2,4-trifluoro-5-methylbenzene
Molecular Weight (g/mol) 146.112
PubChem CID 17965610
Formula Weight 146.11
Percent Purity 98%
Chemical Name or Material 2,4,5-Trifluorotoluene
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.