Learn More
Medchemexpress LLC 2,4,5-trimethoxybenzaldehyde | 4460-86-0 | MFCD00003312 | 99.8% | 196.20 g/mol | C10H12O4 | 100 G
SDP
Catalog No. 5000211780
Encompass_Preferred
Asaraldehyde (2,4,5-trimethoxybenzaldehyde) is a natural small-molecule reported to inhibit cyclooxygenase-2 (COX-2). It is supplied as a research/analytical standard with reported purity 99.83%, molecular weight 196.20 g/mol, and molecular formula C10H12O4. Literature cited by the supplier reports COX-2 inhibition with an IC50 of 100 μg/mL.
  • Acts as a COX-2 inhibitor (reported IC50 100 μg/mL).
  • High reported purity of 99.83%.
  • Molecular weight 196.20 g/mol; formula C10H12O4.
  • Provided as a research standard for biochemical and pharmacological assays.
  • Available in laboratory-scale quantities, including 100 g packaging.
Catalog No. 50-002-11780 Supplier Medchemexpress LLC Supplier No. HY100580100G
Asaraldehyde (2,4,5-trimethoxybenzaldehyde) is a natural small-molecule reported to inhibit cyclooxygenase-2 (COX-2). It is supplied as a research/analytical standard with reported purity 99.83%, molecular weight 196.20 g/mol, and molecular formula C10H12O4. Literature cited by the supplier reports COX-2 inhibition with an IC50 of 100 μg/mL.
  • Acts as a COX-2 inhibitor (reported IC50 100 μg/mL).
  • High reported purity of 99.83%.
  • Molecular weight 196.20 g/mol; formula C10H12O4.
  • Provided as a research standard for biochemical and pharmacological assays.
  • Available in laboratory-scale quantities, including 100 g packaging.