Learn More
Medchemexpress LLC 2,4,5-trimethoxybenzoic acid | 490-64-2 | MFCD00002435 | 100.0% | 212.20 g/mol | C10H12O5 | 50g
SDP
Catalog No. 5000268953
Encompass_Preferred
2 4 5-Trimethoxybenzoic acid (Asaronic acid) is a compound identified in purple perilla extracts 2 4 5-Trimethoxybenzoic acid inhibits LPS (HY-D1056)-induced inflammatory responses inhibits the activation of NF- B and STAT signaling pathways 2 4 5-Trimethoxybenzoic acid inhibits M1 macrophage phenotype-mediated inflammation in diabetes[1]
Catalog No. 50-002-68953 Supplier Medchemexpress LLC Supplier No. HYY058650G
Add to Cart
Add to Cart
2 4 5-Trimethoxybenzoic acid (Asaronic acid) is a compound identified in purple perilla extracts 2 4 5-Trimethoxybenzoic acid inhibits LPS (HY-D1056)-induced inflammatory responses inhibits the activation of NF- B and STAT signaling pathways 2 4 5-Trimethoxybenzoic acid inhibits M1 macrophage phenotype-mediated inflammation in diabetes[1]