Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 97%

Catalog No. AAL1664530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1664530 250 g
AAL1664518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1664530 Supplier Thermo Scientific Chemicals Supplier No. L1664530
  • $355.00
    $285.65 / Each of 1

    Save $69.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2554-06-5 | C9H12O2 | 152.19 g/mol

2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Incompatible with acids, bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2554-06-5
MDL Number MFCD00040293

Specifications

Boiling Point 110°C (10mmHg)
Melting Point -43°C
Quantity 250 g
Flash Point 112°C (233°F)
Refractive Index 1.434
Density 0.997
Molecular Formula C9H12O2
Beilstein 1807272
Synonym phenol, 2-ethoxy-4-methyl, 2-ethoxy-p-cresol, methyl creosol, acmc-20anwy, ethoxy-p-cresol, 2-ethoxy-4-methyl-phenol, phenol,2-ethoxy-4-methyl
Solubility Information Miscible with most organic solvents.
InChI Key ZWQBCCVEOIYNDL-UHFFFAOYSA-N
SMILES CCOC1=C(C=CC(=C1)C)O
IUPAC Name 2-ethoxy-4-methylphenol
Molecular Weight (g/mol) 152.19
PubChem CID 75715
Formula Weight 344.67
Percent Purity 97%
Chemical Name or Material 2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 219-863-1
RTECSNumber GZ4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.