Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tri-tert-butylphenol, TCI America™
SDP

Catalog No. T035925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T035925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T035925G Supplier TCI America Supplier No. T035925G

Chemical Identifiers

CAS 732-26-3
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.44
MDL Number MFCD00008821
InChI Key PFEFOYRSMXVNEL-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylphenol, voidox, alkofen b, 2,4,6-tris tert-butyl phenol, 2,4,6-tri-t-butylphenol, phenol, 2,4,6-tris 1,1-dimethylethyl, phenol, 2,4,6-tri-tert-butyl, 2,4,6-tris 1,1-dimethylethyl phenol, unii-99j2vic675, tris 1,1-dimethylethyl phenol
PubChem CID 12902
IUPAC Name 2,4,6-tri-tert-butylphenol
SMILES CC(C)(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 131°C
Quantity 25 g
UN Number 3077
Formula Weight 262.44
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri-tert-butylphenol
EINECSNumber (3)-0540
RTECSNumber SN3570000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.