Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4,6-Triaminopyrimidine

Catalog No. T4580225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T4580225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T4580225G Supplier Sigma Aldrich Supplier No. T4580225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2α-bromoacetophenone (2-bromoacetophenone) undergoes enantioselective addition reaction with phenylacetylene catalyzed by chiral camphorsulfonamide
  • It reacts with aliphatic primary amines in the presence of palladium catalyst to afford 3-methyleneisoindolin-1-ones.2α-bromoacetophenone (2-bromoacetophenone) was used in the synthesis of novel series of non-condensed 5,5-bicycles

Specifications

CAS 1004-38-2
Melting Point 249°C to 251°C (lit.)
Molecular Formula C4H7N5
Linear Formula C4H7N5
MDL Number MFCD00006100
Quantity 25 g
Synonym 2,4,6-Pyrimidinetriamine
Molecular Weight (g/mol) 125.13
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.