Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromoaniline 98.0+%, TCI America™
SDP

Catalog No. T034525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T034525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T034525G Supplier TCI America Supplier No. T034525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 147-82-0
Melting Point 120°C
Color White
Molecular Formula C6H4Br3N
Quantity 25 g
MDL Number MFCD00007634
Synonym benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline
InChI Key GVPODVKBTHCGFU-UHFFFAOYSA-N
SMILES NC1=C(Br)C=C(Br)C=C1Br
IUPAC Name 2,4,6-tribromoaniline
Molecular Weight (g/mol) 329.82
PubChem CID 8986
Formula Weight 329.82
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromoaniline
Show More Show Less
EINECSNumber (3)-0292
RTECSNumber BZ0175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.