Learn More
Medchemexpress LLC 2,4,6-Tribromophenol-d2 | 1219795-42-2 | 99.9% | 332.81 | 10 MG
SDP
Catalog No. 5000453186
Encompass_Preferred
2,4,6-Tribromophenol-d2 is the deuterium labeled 2,4,6-Tribromophenol. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53186 Supplier Medchemexpress LLC Supplier No. HYW009140S10MG
2,4,6-Tribromophenol-d2 is the deuterium labeled 2,4,6-Tribromophenol. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.