Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromophenol Solution, 2000μg/mL in Methanol, Ultra Scientific

Catalog No. USATS1811
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USATS1811 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. USATS1811 Supplier Agilent Technologies Supplier No. ATS1811
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118-79-6 , 75-09-2
Molecular Formula C6H3Br3O
Molecular Weight (g/mol) 330.80
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
PubChem CID 1483
ChEBI CHEBI:47696
IUPAC Name 2,4,6-tribromophenol
SMILES OC1=C(Br)C=C(Br)C=C1Br

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Chemical Name or Material 2, 4, 6-Tribromophenol, 2000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.