Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromophenol 98.0+%, TCI America™
SDP

Catalog No. T034925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T034925G 25 g
T0349500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. T034925G Supplier TCI America Supplier No. T034925G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 118-79-6
Molecular Formula C6H3Br3O
Molecular Weight (g/mol) 330.80
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Synonym tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
PubChem CID 1483
ChEBI CHEBI:47696
IUPAC Name 2,4,6-tribromophenol
SMILES OC1=C(Br)C=C(Br)C=C1Br

Specifications

Melting Point 94°C
Boiling Point 244°C
Quantity 25 g
UN Number 3077
Formula Weight 330.80
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromophenol
EINECSNumber (3)-0959
RTECSNumber SN1225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.