Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromopyrimidine, Thermo Scientific™

Catalog No. MAY00246DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00246DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00246DA Supplier Thermo Scientific Chemicals Supplier No. MAY00246DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36847-11-7
Molecular Formula C4HBr3N2
Molecular Weight (g/mol) 316.78
MDL Number MFCD00233942
InChI Key AHEYFWKLKMOHCI-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-tribromo, pubchem6896, acmc-1ae09, 2,4,6-tribromopyrimidine
PubChem CID 258580
IUPAC Name 2,4,6-tribromopyrimidine
SMILES BrC1=CC(Br)=NC(Br)=N1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 316.78
Percent Purity 95%
Chemical Name or Material 2, 4, 6-Tribromopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.