Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichloro-5-pyrimidinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. T31001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31001G 1 g
T31005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31001G Supplier TCI America Supplier No. T31001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50270-27-4
Molecular Formula C5HCl3N2O
Molecular Weight (g/mol) 211.426
MDL Number MFCD02257700
InChI Key KVJIRFGNHAAUNQ-UHFFFAOYSA-N
Synonym 2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehyde
PubChem CID 10932746
IUPAC Name 2,4,6-trichloropyrimidine-5-carbaldehyde
SMILES C(=O)C1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 1 g
Formula Weight 211.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trichloro-5-pyrimidinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.