Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T20355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20355G 5 g
T203525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20355G Supplier TCI America Supplier No. T20355G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51527-73-2
Molecular Formula C6H2Cl4O2S
Molecular Weight (g/mol) 279.94
MDL Number MFCD00052973
InChI Key WHJAQKAAIOHCGN-UHFFFAOYSA-N
Synonym 2,4,6-trichlorobenzene-1-sulfonyl chloride, 2,4,6-trichlorophenylsulfonyl chloride, 2,4,6-trichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-trichloro, 2,4,6-trichlorobenzenesulfonylchloride, pubchem5175, acmc-209ku8, ksc269e6l, chloro 2,4,6-trichlorophenyl sulfone, 2,4,6 trichlorobenzenesulfonyl chloride
PubChem CID 521335
IUPAC Name 2,4,6-trichlorobenzenesulfonyl chloride
SMILES C1=C(C=C(C(=C1Cl)S(=O)(=O)Cl)Cl)Cl

Specifications

Melting Point 49°C
Quantity 5 g
UN Number 3261
Formula Weight 279.94
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trichlorobenzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.