Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
88-06-2
C6H3Cl3O
197.45
MFCD00002172
LINPIYWFGCPVIE-UHFFFAOYSA-N
dowicide 2s,phenachlor,phenol, 2,4,6-trichloro,omal,phenaclor,rcra waste number u231,dowcide 2s,1,3,5-trichloro-2-hydroxybenzene,2,4,6-trichlorfenol,unii-mhs8c5bauz
6914
CHEBI:28755
2,4,6-trichlorophenol
C1=C(C=C(C(=C1Cl)O)Cl)Cl
88-06-2
C6H3Cl3O
197.45
MFCD00002172
LINPIYWFGCPVIE-UHFFFAOYSA-N
dowicide 2s,phenachlor,phenol, 2,4,6-trichloro,omal,phenaclor,rcra waste number u231,dowcide 2s,1,3,5-trichloro-2-hydroxybenzene,2,4,6-trichlorfenol,unii-mhs8c5bauz
6914
CHEBI:28755
2,4,6-trichlorophenol
C1=C(C=C(C(=C1Cl)O)Cl)Cl
Melting Point 64.0°C to 68.0°C
Infrared Spectrum Authentic
Flash Point 99°C
Color Beige to Brown
Quantity 100g
Boiling Point 246.0°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl3C6H2OH
Beilstein 06, 190
Merck Index 15, 9809
Solubility Information Solubility in water: 0.8g/L. Other solubilities: soluble in aromatic solvents and caustic soda sol.,soluble in (g/100g) acetone 525; benzene 113;,ccl4 37; diacetone alcohol 335; ether 354;,methanol 525; pine oil 163; toluene 100;,turpentine 37
Formula Weight 197.45
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material 2, 4, 6-Trichlorophenol
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger