Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4,6-Trichloropyridine | 16063-69-7 | 182.44 | 10 G
SDP

Catalog No. 5000445359
Encompass_Preferred

2,4,6-Trichloropyridine is a biochemical reagent suitable for life science-related research. This organic compound can be utilized as a biological material and is intended for research use only.

  • Purity: 99.98%
  • Molecular weight: 182.44
  • Appearance: White to light yellow
  • Physical state: Solid below 33°C, liquid above 37°C
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-45359 Supplier Medchemexpress LLC Supplier No. HYW00271810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,6-Trichloropyridine is a biochemical reagent suitable for life science-related research. This organic compound can be utilized as a biological material and is intended for research use only.

  • Purity: 99.98%
  • Molecular weight: 182.44
  • Appearance: White to light yellow
  • Physical state: Solid below 33°C, liquid above 37°C
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.