Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. T19085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19085G Supplier TCI America Supplier No. T19085G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96606-37-0
Molecular Formula C7H2F3N
Molecular Weight (g/mol) 157.095
MDL Number MFCD00042399
InChI Key HTKFGTCCOJIUIK-UHFFFAOYSA-N
Synonym benzonitrile, 2,4,6-trifluoro, 2,4,6-trifluorobenzenecarbonitrile, ncr bf df ff, pubchem1565, acmc-209s7e, ksc488g9f, 2,4,6-trifluoro-benzonitrile, 2,4,6-trifluorobenzonitrile, benzonitrile, 2,4,6-trifluoro-, radical ion 1-9ci
PubChem CID 737177
IUPAC Name 2,4,6-trifluorobenzonitrile
SMILES C1=C(C=C(C(=C1F)C#N)F)F

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 157.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trifluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.