Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluorobenzotrifluoride, 96%

Catalog No. AAL1816706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1816706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1816706 Supplier Thermo Scientific Chemicals Supplier No. L1816706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 122030-04-0 | C7H2F6 | 200.083 g/mol

2,4,6-Trifluorobenzotrifluoride finds its application as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluorobenzotrifluoride finds its application as an intermediate in organic syntheses.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122030-04-0
Molecular Formula C7H2F6
Molecular Weight (g/mol) 200.083
MDL Number MFCD00190109
InChI Key SGPXFVDJRSZIPK-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzotrifluoride, 1,3,5-trifluoro-2-trifluoromethyl benzene, pubchem4877, 2,4,6-trifluorobenzotrifluorode, 2,4,6-tri-fluorobenzotrifluoride, benzene, 1,3,5-trifluoro-2-trifluoromethyl, alpha,alpha,alpha,2,4,6-hexafluorotoluene, alpha,alpha,alpha,2,4,6-hexafluorotoluene;1,3,5-trifluoro-2-trifluoromethyl benzene
PubChem CID 2734030
IUPAC Name 1,3,5-trifluoro-2-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1F)C(F)(F)F)F)F

Specifications

Density 1.475
Boiling Point 105°C to 107°C
Flash Point 22°C (71°F)
Refractive Index 1.385
Quantity 5 g
UN Number UN1993
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 200.08
Percent Purity 99%
Chemical Name or Material 2,4,6-Trifluorobenzotrifluoride
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.