Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluorobenzoyl chloride, 97%

Catalog No. AAB2032303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2032303 1 g
AAB2032306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2032303 Supplier Thermo Scientific Chemicals Supplier No. B2032303
  • $63.25
    $53.65 / Each of 1

    Save $9.60

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 79538-29-7 | C7H2ClF3O | 194.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79538-29-7
Molecular Formula C7H2ClF3O
Molecular Weight (g/mol) 194.537
MDL Number MFCD00070810
InChI Key SIFIJQFBERMWMU-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci
PubChem CID 144895
IUPAC Name 2,4,6-trifluorobenzoyl chloride
SMILES C1=C(C=C(C(=C1F)C(=O)Cl)F)F

Specifications

Density 1.499
Boiling Point 167°C to 168°C
Refractive Index 1.484
Quantity 1 g
UN Number UN1760
Beilstein 1958563
Sensitivity Moisture sensitive
Formula Weight 194.54
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzoyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.