Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T28045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2804-5G 5 g
T28041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2804-5G Supplier TCI America Supplier No. T28045G
Add to Cart
Add to Cart

Specifications

CAS 182482-25-3
Melting Point 235°C
Molecular Formula C6H4BF3O2
Quantity 5 g
MDL Number MFCD01863169
Synonym 2,4,6-trifluorophenyl boronic acid, 2,4,6-trifluorobenzeneboronic acid, 2,4,6-trifluorophenylboronicacid, 2,4,6-trifluorophenyl boranediol, boronic acid, 2,4,6-trifluorophenyl
InChI Key IPEIGKHHSZFAEW-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1F)F)F)(O)O
IUPAC Name (2,4,6-trifluorophenyl)boronic acid
Molecular Weight (g/mol) 175.901
PubChem CID 2779329
Formula Weight 175.90
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.