Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluoropyridine, 97%

Catalog No. AAL1954114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1954114 25 g
AAL1954106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1954114 Supplier Thermo Scientific Chemicals Supplier No. L1954114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3512-17-2 | C5H2F3N | 133.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3512-17-2
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001161
InChI Key UZDRWXKBKVVUTE-UHFFFAOYSA-N
Synonym 2,4,6-trifluoro-pyridine, pyridine, 2,4,6-trifluoro, pubchem7794, 2,4,6 trifluoropyridine, acmc-1cp7f, 2,4,6-tris fluoranyl pyridine
PubChem CID 2783289
IUPAC Name 2,4,6-trifluoropyridine
SMILES C1=C(C=C(N=C1F)F)F

Specifications

Density 1.382
Boiling Point 102°C
Flash Point 17°C (62°F)
Refractive Index 1.413
Quantity 25 g
UN Number UN2924
Beilstein 1364338
Formula Weight 133.07
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluoropyridine
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.