Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2',4',6'-Trihydroxyacetophenone monohydrate

Catalog No. T6460225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T6460225G 25 g
T6460210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T6460225G Supplier Sigma Aldrich Supplier No. T6460225G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2',4',6'-Trihydroxyacetophenone monohydrate

Specifications

CAS 249278-28-2
Melting Point 219°C to 221°C (lit.)
Molecular Formula C8H8O4 -+ H2O
Linear Formula (HO)3C6H2COCH3-+H2O
MDL Number MFCD00149091
Quantity 25 g
Synonym 2-Acetylphloroglucinol; THAP
Molecular Weight (g/mol) 186.16
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.