Learn More
Medchemexpress LLC 2,4,6-Trihydroxybenzaldehyde | 487-70-7 | 154.12 | 25 G
SDP
Catalog No. 5000449299
Encompass_Preferred
2,4,6-Trihydroxybenzaldehyde is an orally active NF-κB inhibitor that demonstrates anti-tumor, anti-cancer cell proliferative, and anti-obesity activities. It inhibits adipocyte differentiation and lipid accumulation in 3T3-L1 cells and down-regulates PPARγ, C/EBPα, SREBP-1c, and FAS protein expression levels. In vivo, oral administration reduces high-fat diet-induced weight gain and lowers serum levels of glucose, triglycerides, and total cholesterol.
  • Inhibits adipocyte differentiation
  • Reduces lipid accumulation
  • Down-regulates PPARγ, C/EBPα, SREBP-1c, and FAS protein expression
  • Reduces high-fat diet-induced weight gain
  • Lowers serum levels of glucose, triglycerides, and total cholesterol
Catalog No. 50-004-49299 Supplier Medchemexpress LLC Supplier No. 25GHYW005130
2,4,6-Trihydroxybenzaldehyde is an orally active NF-κB inhibitor that demonstrates anti-tumor, anti-cancer cell proliferative, and anti-obesity activities. It inhibits adipocyte differentiation and lipid accumulation in 3T3-L1 cells and down-regulates PPARγ, C/EBPα, SREBP-1c, and FAS protein expression levels. In vivo, oral administration reduces high-fat diet-induced weight gain and lowers serum levels of glucose, triglycerides, and total cholesterol.
  • Inhibits adipocyte differentiation
  • Reduces lipid accumulation
  • Down-regulates PPARγ, C/EBPα, SREBP-1c, and FAS protein expression
  • Reduces high-fat diet-induced weight gain
  • Lowers serum levels of glucose, triglycerides, and total cholesterol