Learn More
Apexbio Technology LLC 2,4,6-Triiodophenol 609-23-4 25mg
SDP
Catalog No. 5000096283
Encompass_Preferred
2 4 6-Triiodophenol (CAS 609-23-4) is a small molecule inhibitor known to block the biosynthetic pathway of leukotriene B4 (LTB4) thereby modulating the production of pro-inflammatory mediators By targeting LTB4 synthesis it serves as a valuable tool for studying inflammatory processes and related signaling mechanisms Additionally this compound has been reported to induce apoptosis in murine blastocyst-stage cells making it relevant for investigations into early embryonic development embryotoxicity and the regulation of apoptotic pathways
Catalog No. 50-000-96283 Supplier Apexbio Technology LLC Supplier No. A470125
2 4 6-Triiodophenol (CAS 609-23-4) is a small molecule inhibitor known to block the biosynthetic pathway of leukotriene B4 (LTB4) thereby modulating the production of pro-inflammatory mediators By targeting LTB4 synthesis it serves as a valuable tool for studying inflammatory processes and related signaling mechanisms Additionally this compound has been reported to induce apoptosis in murine blastocyst-stage cells making it relevant for investigations into early embryonic development embryotoxicity and the regulation of apoptotic pathways