Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4,6-triiodophenol | 609-23-4 | MFCD00002179 | 99.5% | 471.80 g/mol | C6H3I3O | 100 MG
SDP

Catalog No. 5000210337
Encompass_Preferred

2,4,6-triiodophenol is an iodinated phenol supplied for laboratory research and described as an orally active inhibitor of leukotriene B4 (LTB4) synthesis. It is provided as a solid and has documented biological activity including induction of apoptosis in mouse blastocysts.

  • cas number: 609-23-4.
  • chemical formula: C6H3I3O.
  • molecular weight: 471.80 g/mol.
  • reported purity: catalog lists 98.0%; batch COA reports 99.52%.
  • biological activity: LTB4 synthesis inhibitor.
  • observed effect: can induce apoptosis in mouse blastocysts.
  • form and packaging: solid powder, supplied in multiple sizes including 100 mg.

Catalog No. 50-002-10337 Supplier Medchemexpress LLC Supplier No. HYW073687100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,6-triiodophenol is an iodinated phenol supplied for laboratory research and described as an orally active inhibitor of leukotriene B4 (LTB4) synthesis. It is provided as a solid and has documented biological activity including induction of apoptosis in mouse blastocysts.

  • cas number: 609-23-4.
  • chemical formula: C6H3I3O.
  • molecular weight: 471.80 g/mol.
  • reported purity: catalog lists 98.0%; batch COA reports 99.52%.
  • biological activity: LTB4 synthesis inhibitor.
  • observed effect: can induce apoptosis in mouse blastocysts.
  • form and packaging: solid powder, supplied in multiple sizes including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.