Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triisopropylbenzenesulfonyl Azide (wetted with ca. 10% Water) (unit weight on dry weight basis) 98.0+%, TCI America™
SDP

Catalog No. T34345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34345G 5 g
T34341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34345G Supplier TCI America Supplier No. T34345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36982-84-0
Molecular Formula C15H23N3O2S
Molecular Weight (g/mol) 309.428
MDL Number MFCD00859286
InChI Key AEMWUHCKKDPRSK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide, trisyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide,, benzenesulfonyl azide, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropyl-benzenesulfonyl azide, n-diazo-2,4,6-triisopropyl-benzenesulfonamide, 2,4,6-tris 1-methylethyl-benzenesulfonyl azide, 2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water, trisyl azide solution
PubChem CID 371707
IUPAC Name N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
Formula Weight 309.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Azide (wetted with ca. 10% Water) (unit weight on dry weight basis
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.