Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethoxybenzaldehyde, 98%

Catalog No. AC160530100
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160530100 10 g Glass bottle
AC160530250 25 g Glass bottle
AC160532500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160530100 Supplier Thermo Scientific Chemicals Supplier No. 160530100
Add to Cart

CAS: 830-79-5 | C10H12O4 | 196.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-79-5
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.2
MDL Number MFCD00003313
InChI Key CRBZVDLXAIFERF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
PubChem CID 70019
IUPAC Name 2,4,6-trimethoxybenzaldehyde
SMILES COC1=CC(=C(C(=C1)OC)C=O)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 118.0°C to 120.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CHO
Quantity 10 g
Beilstein 08,39
Formula Weight 196.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trimethoxybenzaldehyde, 98%
Show More Show Less
EINECSNumber 212-598-2
RTECSNumber CU8460200
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.