Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2,4,6-trimethyl-phenoxy)-acetic acid | 13333-81-8 | MFCD00224887 | 99.5% | 194.23 g/mol | C11H14O3 | 10 G
SDP

Catalog No. 5000281670
Encompass_Preferred

(2,4,6-Trimethyl-phenoxy)-acetic acid is an herbicide-related analytical reference standard used for method development, quality control, and research workflows that require a characterized small-molecule standard. It is supplied as a powder in laboratory-scale quantities and is accompanied by product documentation for traceability and handling guidance.

  • High purity suitable for analytical use (≈99.5%).
  • Provided as a powder for laboratory workflows.
  • Useful for method development and quality control applications.
  • Contains CAS number 13333-81-8 for unambiguous identification.
  • Molecular formula C11H14O3 and molecular weight 194.23 g/mol.

Catalog No. 50-002-81670 Supplier Medchemexpress LLC Supplier No. HYW26430610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2,4,6-Trimethyl-phenoxy)-acetic acid is an herbicide-related analytical reference standard used for method development, quality control, and research workflows that require a characterized small-molecule standard. It is supplied as a powder in laboratory-scale quantities and is accompanied by product documentation for traceability and handling guidance.

  • High purity suitable for analytical use (≈99.5%).
  • Provided as a powder for laboratory workflows.
  • Useful for method development and quality control applications.
  • Contains CAS number 13333-81-8 for unambiguous identification.
  • Molecular formula C11H14O3 and molecular weight 194.23 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.